Learn More
Bohlender™ SICCO™ Glove Box Pure with Transfer Chamber
Glove Box Pure with Transfer Chamber Aluminum frame with two glove ports and one pair of gloves.
Brand:  Bohlender™ V1740-08
Product Code. 17107684
  • £12200.00 / Each
Explore more special offers
Description
Description
  • Aluminum frame with panels made of acrylic glass
  • Base panel made of polyethylene
  • Front panel with two glove ports and one pair of gloves size 9.75 made of UV-resistant EPDM
  • Transfer chamber with two doors on the right, external door with latch to prevent accidental opening
  • Tray extends into workspace with opening of the internal door for easy removal of inserted products
  • Integrated ultraviolet lamp with life-time monitoring on the top side for air and surface disinfection in the workspace
  • Exchangeable lamp
  • Ultraviolet lamp is an ozone-free radiation source that does not emit any radiation below 220 nm
  • Robust and light-weight aluminum construction
  • Superb all-round visibility
  • Non-slip rubber feet
  • Cable lead-ins for electric devices
  • Large front opening
Specifications
Specifications
Acrylic Glass, Polyethylene
600 mm
700 mm
230 V
48.5 lb.
48 in.
24 in.
28 in.
50 Hz
290 L
22 kg
1210 mm
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Bohlender™ SICCO™ Glove Box Pure with Transfer Chamber > Glove Box Pure with Transfer Chamber; Material: Acrylic Glass, Polyethylene; Voltage: 230 V; Depth (Metric) Exterior: 600 mm; Height (Metric) Exterior: 700 mm; Width (Metric) Exterior: 1210 mm