missing translation for 'onlineSavingsMsg'
Learn More

Epredia™ Signature Series™ Paraffin

Minimize compression and obtain rapid infiltration with this unique blend of polymers and highly refined paraffin with narrow carbon chain distribution.

£105.00 - £120.00

Specifications

Melting Point 54° to 57°C
Quantity 2 lb. bag (0.9kg)
View More Specs

Products 2
Product Code Brand Product Type Price Quantity & Availability  
Product Code Brand Product Type Price Quantity & Availability  
12086669
View Documents
Epredia™
8338
Paraffin, Type H N/A N/A N/A
12687766
View Documents
Epredia™
8339
Paraffin, Type L
£120.00
Pack of 10
Please to purchase this item. Need a web account? Register with us today!
Description

Description

  • Triple-filtered to eliminate impurities
  • Type H allows for ultra-thin sections
  • Type L may reduce infiltration times during processing
  • Both Type L and Type H can be used for infiltration and embedding, and have a melting point of 55° to 57°C

Ordering Information: Packaged in easy-to-open, resealable bags.

Specifications

Specifications

54° to 57°C
2 lb. bag (0.9kg)
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Epredia™ Signature Series™ Paraffin

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.