Learn More
Silicon carbide, beta-phase, nanopowder, 95% min, Thermo Scientific Chemicals
Description
Silicon carbide is used in applications requiring high endurance, such as car brakes, car clutches and ceramic plates in bulletproof vests. Used in electronic applications, measure gas temperatures in an optical technique called thin filament pyrometry. Used in jewelry, steel production, grapheme production. It is also used in production of exponential resistors, wire sawing, construction industry, a filler in plastics and composites, ceramic sintered parts, ferrites and soft metals.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Specifications
Melting Point | 2700°C |
Color | Black to Gray |
Physical Form | Powder |
Quantity | 5 g |
Merck Index | 14,8492 |
Solubility Information | Insoluble in water, alcohol; Soluble in fused alkali |
IUPAC Name | methanidylidynesilanylium |
Formula Weight | 40.1 |
Percent Purity | ≥95% |
Odor | Odorless |
Show More |
Product Suggestions
Customers who viewed this item also viewed.
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
Y28I028 | Certificate of Analysis | 17/01/2023 | 043884.14,043884.06,043884.22 |
S06I004 | Certificate of Analysis | 04/07/2022 | 043884.14,043884.06,043884.22 |
E24R048 | Certificate of Analysis | 18/11/2021 | 043884.14,043884.06,043884.22 |
B27N28 | Certificate of Analysis | 18/11/2021 | 043884.14,043884.06,043884.22 |
K10P17 | Certificate of Analysis | 18/11/2021 | 043884.14,043884.06,043884.22 |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.