missing translation for 'onlineSavingsMsg'
Learn More
Silver benzoate, 99%, Thermo Scientific Chemicals Product Code.: 10328350

Silver benzoate, 99%, Thermo Scientific Chemicals

Product Code. 10328350
10 g, Glass bottle
Click to view available options
Quantity:
10 g
50 g
Packaging:
Glass bottle
This item is not returnable. View return policy

Product Code. 10328350

Brand: Thermo Scientific Acros 210770100

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 532-31-0
Molecular Formula C7H5AgO2
Molecular Weight (g/mol) 228.98
MDL Number MFCD00013030
InChI Key CLDWGXZGFUNWKB-UHFFFAOYSA-M
Synonym silver benzoate hydrate
PubChem CID 87384783
IUPAC Name benzoic acid;silver;hydrate
SMILES [Ag+].[O-]C(=O)C1=CC=CC=C1

Specifications

Color White to Beige
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Linear Formula C6H5CO2Ag
Quantity 10 g
Merck Index 15, 1093
Solubility Information Solubility in water: 0.26%. Other solubilities: slightly soluble in alcohol
Formula Weight 228.99
Percent Purity 99%
Physical Form Powder
Chemical Name or Material Silver benzoate, 99%
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

2 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0469040 Certificate TypeCertificate of Analysis Date31/01/2025 Catalog Number210770100,210770500
Lot NumberA0409619 Certificate TypeCertificate of Analysis Date20/12/2023 Catalog Number210770100,210770500
Product Identifier
  • Silver benzoate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Silver benzoate, 99%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.