Learn More
Silver sulfate, 99%, extra pure, Thermo Scientific Chemicals
£57.49 - £1365.80
Chemical Identifiers
CAS | 10294-26-5 |
---|---|
Molecular Formula | Ag2H2O4S |
Molecular Weight (g/mol) | 313.81 |
MDL Number | MFCD00003407 |
InChI Key | YPNVIBVEFVRZPJ-UHFFFAOYSA-N |
Synonym | silver sulfate, disilver sulfate, disilver 1+ sulfate, unii-8qg6hv4zpo, disilver 1+ sulphate, sulfuric acid, disilver 1+ salt, 8qg6hv4zpo, disilver 1+ ion sulfate, sulfuric acid, silver 1+ salt 1:2, sulfuric acid disilver i salt |
PubChem CID | 159865 |
IUPAC Name | disilver(1+) sulfuric acid |
SMILES | [Ag+].[Ag+].OS(O)(=O)=O |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10174930
|
Thermo Scientific Acros
209470100 |
10 g | Glass bottle |
£57.49
10g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10606622
|
Thermo Scientific Acros
209471000 |
100 g | Glass bottle |
£331.65
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10783884
|
Thermo Scientific Acros
209475000 |
500 g | Glass bottle |
£1365.80
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
10294-26-5 | |
313.81 | |
YPNVIBVEFVRZPJ-UHFFFAOYSA-N | |
159865 | |
[Ag+].[Ag+].OS(O)(=O)=O |
Ag2H2O4S | |
MFCD00003407 | |
silver sulfate, disilver sulfate, disilver 1+ sulfate, unii-8qg6hv4zpo, disilver 1+ sulphate, sulfuric acid, disilver 1+ salt, 8qg6hv4zpo, disilver 1+ ion sulfate, sulfuric acid, silver 1+ salt 1:2, sulfuric acid disilver i salt | |
disilver(1+) sulfuric acid |
Specifications
1085.0°C | |
10294-26-5 | |
Fine Crystalline Powder | |
Ag2H2O4S | |
MFCD00003407 | |
15, 8668 | |
Solubility in water: 7.4g/L (20°C). Other solubilities: soluble in nh4oh, nitric acid and sulfuric acid, insoluble in alcohol | |
[Ag+].[Ag+].OS(O)(=O)=O | |
313.81 | |
311.79 | |
Extra Pure | |
Silver sulfate |
652.0°C | |
White | |
98.5% min. (Argentometry) | |
Ag2SO4 | |
01,1015; 03,254; 04,435 | |
silver sulfate, disilver sulfate, disilver 1+ sulfate, unii-8qg6hv4zpo, disilver 1+ sulphate, sulfuric acid, disilver 1+ salt, 8qg6hv4zpo, disilver 1+ ion sulfate, sulfuric acid, silver 1+ salt 1:2, sulfuric acid disilver i salt | |
YPNVIBVEFVRZPJ-UHFFFAOYSA-N | |
disilver(1+) sulfuric acid | |
159865 | |
99% | |
1.84g/mL |
Safety and Handling
GHS H Statement
Causes serious eye damage.
GHS P Statement
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Immediately call a POISON CENTER or doctor/physici
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 233-653-7
- Silver sulfate
Signal Word
- Danger
Hazard Category
- LONG-TERM AQUATIC HAZARD Chronic 1
- ACUTE AQUATIC Acute 1
- Serious eye damage/eye irritation Category 1
Hazard Statement
- H318-Causes serious eye damage.
- H400-Very toxic to aquatic life.
- H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
- MIXTURE LIST-Contains : Silver sulfate
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.