missing translation for 'onlineSavingsMsg'
Learn More
Thermo Scientific Chemicals Sirtinol Product Code.: 15476419

Thermo Scientific Chemicals Sirtinol

Product Code. 15476419
10 mg
Click to view available options
Quantity:
10 mg
This item is not returnable. View return policy

Product Code. 15476419

Brand: Thermo Scientific Chemicals J64486.MA

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

 

Sirtinol is a cell permeable specific inhibitor of sirtuin NAD-dependant deacetylases

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

Quantity 10 mg
Description Off-white to Yellow
Content And Storage -30°C to -10°C
For Use With (Application) Sirtinol is a cell permeable specific inhibitor of sirtuin NAD-dependant deacetylases
CAS 410536-97-9
Molecular Formula C26H22N2O2, C26H22N2O2
Formula Weight 394.46g/mol
Synonym (E)-2-((2-Hydroxynaphthalen-1-yl)methyleneamino)-N-(1-phenylethyl)benzamide
Solubility Soluble in water, DMSO and methanol
Product Type Signal Transduction Reagent
Form Crystals
InChI Key YUGODMKHHCZZOI-ZVTCDHROSA-N
SMILES C[C@H](NC(=O)C1=CC=CC=C1N\C=C1/C(=O)C=CC2=CC=CC=C12)C1=CC=CC=C1
IUPAC Name 2-({[(1Z)-2-oxo-1,2-dihydronaphthalen-1-ylidene]methyl}amino)-N-[(1S)-1-phenylethyl]benzamide
Molecular Weight (g/mol) 394.47
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberR06K015 Certificate TypeCertificate of Analysis Date14/05/2024 Catalog NumberJ64486.MA
Lot NumberZ19J005 Certificate TypeCertificate of Analysis Date12/01/2024 Catalog NumberJ64486.MA
Lot NumberS16J035 Certificate TypeCertificate of Analysis Date26/06/2023 Catalog NumberJ64486,J64486.MA
Lot NumberP27J021 Certificate TypeCertificate of Analysis Date09/05/2023 Catalog NumberJ64486.MA
Lot NumberM24I021 Certificate TypeCertificate of Analysis Date17/11/2022 Catalog NumberJ64486.MA

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific Chemicals Sirtinol >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.