missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Asecos™ SLX-CLASSIC Recirculating Air Safety Storage Cabinets
For the safe and approved storage of aggressive, non-flammable hazardous materials in workrooms.
POA
Specifications
Certifications/Compliance | EN 14727 |
---|---|
Color | Light Gray |
Material | Steel |
Product Type | Safety storage cabinet |
Door Style | Wing |
Product Code | Brand | Load Bearing Capacity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Load Bearing Capacity | Price | Quantity & Availability | |||||
15527839
|
Asecos™
3293600130607 |
8 x Shelves with sump 25 kg/11 L each, Maximum load 300 kg |
POA
Each |
N/A | Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15695677
|
Asecos™
3293600130608 |
12 x Shelves with sump: 25 kg/11 L each, Maximum load: 300 kg |
POA
Each |
N/A | Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15904690
|
Asecos™
3293900130615 |
6 x Shelves with sump: 25 kg/11 L each, Maximum load: 300 kg |
POA
Each |
N/A | Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
- Robust construction and longevity – cabinet body made of powder-coated sheet steel with safety elements assembled outside the storage compartment for increased protection against corrosion
- Two-in-one – two hermetically separated storage compartments allow the isolated storage of acids and bases in one cabinet
- No unauthorized use – doors lockable with profile cylinder with integration in an existing locking system possible
- Easy alignment – adjusting aids to compensate for uneven floors
- Recirculating air filter system, ideal for the direct installation in working areas with very low noise, only approx. 39 dB
- With multi-stage wideband filter
- Monitoring electronics for extraction include cool-device cable and power plug
- Optical and acoustic alarm including potential-free alarm contact
- Display with innovative touch panel
- Manual check of the filter saturation
- Eco-friendly with longer filter lifetimes and a reusable filter system
Specifications
EN 14727 | |
Steel | |
Wing | |
2 |
Light Gray | |
Safety storage cabinet | |
Storage of aggressive, non-flammable hazardous materials | |
Powder-coated |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Asecos™ SLX-CLASSIC Recirculating Air Safety Storage Cabinets