Learn More
Asecos™ SLX-CLASSIC Recirculating Air Safety Storage Cabinets
For the safe and approved storage of aggressive, non-flammable hazardous materials in workrooms.
Brand:  Asecos™ 3293600130607
 View more versions of this product
Product Code. 15527839
  • £6575.00 / Each
Explore more special offers
Description
Description
  • Robust construction and longevity – cabinet body made of powder-coated sheet steel with safety elements assembled outside the storage compartment for increased protection against corrosion
  • Two-in-one – two hermetically separated storage compartments allow the isolated storage of acids and bases in one cabinet
  • No unauthorized use – doors lockable with profile cylinder with integration in an existing locking system possible
  • Easy alignment – adjusting aids to compensate for uneven floors
  • Recirculating air filter system, ideal for the direct installation in working areas with very low noise, only approx. 39 dB
  • With multi-stage wideband filter
  • Monitoring electronics for extraction include cool-device cable and power plug
  • Optical and acoustic alarm including potential-free alarm contact
  • Display with innovative touch panel
  • Manual check of the filter saturation
  • Eco-friendly with longer filter lifetimes and a reusable filter system
Specifications
Specifications
EN 14727
565 x 485 x 1865 mm
Wing
8 x Shelves with sump 25 kg/11 L each, Maximum load 300 kg
Powder-coated
Light Gray
Safety storage cabinet
8 x 25 kg/11 L PP pull-out shelves with PP sumps
615 x 1197 x 2300 mm
Storage of aggressive, non-flammable hazardous materials
2
330.7 kg
Steel
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Asecos™ SLX-CLASSIC Recirculating Air Safety Storage Cabinets > 8x 25 kg/11 L PP pull-out shelves with PP sumps