Learn More
BRAND™ Soda-Lime Glass Watch Glasses
Stress-reduced to minimize risk of shattering.
£29.30 - £119.00
Product Code | Brand | Diameter (Metric) | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Diameter (Metric) | Price | Quantity & Availability | ||||||
10771572
|
BRAND™
150065 |
125 mm |
£71.40
Pack of 10 |
Estimated Shipment: 13-05-2025
Log in to see stock. |
||||||
10313031
|
BRAND™
150070 |
150 mm |
£71.05
Pack of 10 |
Estimated Shipment: 13-05-2025
Log in to see stock. |
||||||
10079980
|
BRAND™
150020 |
60 mm |
£42.80
Pack of 10 |
In Stock
Log in to see stock. ![]() |
||||||
10273391
|
BRAND™
150045 |
90 mm |
£51.35
Pack of 10 |
Estimated Shipment: 16-05-2025
Log in to see stock. |
||||||
10588831
|
BRAND™
150080 |
200 mm |
£119.00
Pack of 10 |
In Stock
Log in to see stock. |
||||||
10558401
|
BRAND™
150030 |
70 mm |
£37.20
Pack of 10 |
In Stock
Log in to see stock. |
||||||
10389901
|
BRAND™
150050 |
100 mm |
£47.55
Pack of 10 |
In Stock
Log in to see stock. ![]() |
||||||
10455771
|
BRAND™
150040 |
80 mm |
£44.75
Pack of 10 |
In Stock
Log in to see stock. ![]() |
||||||
15201489
|
BRAND™
150015 |
50 mm |
£29.30
Pack of 10 |
In Stock
Log in to see stock. |
||||||
Description
- Soda-lime glass to DIN 12341
- Ground edges
- Stress-reduced to minimize risk of shattering
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Product Title
BRAND™ Soda-Lime Glass Watch Glasses