missing translation for 'onlineSavingsMsg'
Learn More

BRAND™ Soda-Lime Glass Watch Glasses

Stress-reduced to minimize risk of shattering.

Brand:  BRAND™ 150045

32.00 GBP valid until 2025-12-31
Use promo code "25339" to get your promotional price.


 View more versions of this product

Product Code. 10273391

  • £51.35 / Pack of 10
Estimated Shipment: 16-05-2025
to see stock.
Explore more special offers
Add to basket

Description

Description

  • Soda-lime glass to DIN 12341
  • Ground edges
  • Stress-reduced to minimize risk of shattering
TRUSTED_SUSTAINABILITY
Specifications

Specifications

DIN 12341
Soda Lime Glass
Watch Glass
90 mm
Flat
Round
Polished
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

One moment while we fetch your results.
Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
BRAND™ Soda-Lime Glass Watch Glasses > Diameter: 90mm

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.