missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Carbonate, Anhydrous, ULTREX™ Ultrapure Reagent, J.T. Baker™
£585.00 - £1575.00
Chemical Identifiers
CAS | 497-19-8 |
---|---|
Molecular Formula | CNa2O3 |
Molecular Weight (g/mol) | 105.99 |
MDL Number | MFCD00003494 |
InChI Key | CDBYLPFSWZWCQE-UHFFFAOYSA-L |
Synonym | sodium carbonate, disodium carbonate, soda ash, carbonic acid disodium salt, calcined soda, sodium carbonate, anhydrous, carbonic acid, disodium salt, washing soda, sodiumcarbonate, solvay soda |
PubChem CID | 10340 |
ChEBI | CHEBI:29377 |
IUPAC Name | disodium;carbonate |
SMILES | [Na+].[Na+].[O-]C([O-])=O |
Chemical Identifiers
497-19-8 | |
105.99 | |
CDBYLPFSWZWCQE-UHFFFAOYSA-L | |
10340 | |
disodium;carbonate |
CNa2O3 | |
MFCD00003494 | |
sodium carbonate, disodium carbonate, soda ash, carbonic acid disodium salt, calcined soda, sodium carbonate, anhydrous, carbonic acid, disodium salt, washing soda, sodiumcarbonate, solvay soda | |
CHEBI:29377 | |
[Na+].[Na+].[O-]C([O-])=O |
Specifications
497-19-8 | |
MFCD00003494 | |
CDBYLPFSWZWCQE-UHFFFAOYSA-L | |
disodium;carbonate | |
10340 | |
105.99 | |
1L = 2.53kg |
CNa2O3 | |
sodium carbonate, disodium carbonate, soda ash, carbonic acid disodium salt, calcined soda, sodium carbonate, anhydrous, carbonic acid, disodium salt, washing soda, sodiumcarbonate, solvay soda | |
[Na+].[Na+].[O-]C([O-])=O | |
105.99 | |
CHEBI:29377 | |
ULTREX™ Ultrapure Reagent | |
Sodium Carbonate, Anhydrous |