Learn More
Sodium hexafluorophosphate, 99+%, Thermo Scientific Chemicals
£91.60 - £291.00
Chemical Identifiers
CAS | 21324-39-0 |
---|---|
Molecular Formula | F6NaP |
Molecular Weight (g/mol) | 167.95 |
MDL Number | MFCD00011122 |
InChI Key | KMADQUOCJBLXRP-UHFFFAOYSA-N |
Synonym | sodium hexafluorophosphate, sodium hexa-fluoro phosphate, sodium hexafluorophosphate v, napf6, sodium hexaflurophosphate, acmc-1cao2, sodium hexafluoro-, e5-phosphanuide, sodium hexafluoro-$l^ 5-phosphanuide, phosphate 1-, hexafluoro-, sodium 1:1 |
PubChem CID | 5147921 |
IUPAC Name | sodium;hexafluorophosphate |
SMILES | [Na+].F[P-](F)(F)(F)(F)F |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11342158
|
Thermo Scientific Alfa Aesar
013064.14 |
25 g |
£91.60
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11352158
|
Thermo Scientific Alfa Aesar
013064.22 |
100 g |
£291.00
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Sodium hexafluorophosphate is used as an ion pairing agent for the determination of cobalt(II) in water samples. It is used as an important raw material in the preparation of 2-chloro-1,3-bis(dimethylamino)trimethinium hexafluorophosphate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
21324-39-0 | |
167.95 | |
KMADQUOCJBLXRP-UHFFFAOYSA-N | |
5147921 | |
[Na+].F[P-](F)(F)(F)(F)F |
F6NaP | |
MFCD00011122 | |
sodium hexafluorophosphate, sodium hexa-fluoro phosphate, sodium hexafluorophosphate v, napf6, sodium hexaflurophosphate, acmc-1cao2, sodium hexafluoro-, e5-phosphanuide, sodium hexafluoro-$l^ 5-phosphanuide, phosphate 1-, hexafluoro-, sodium 1:1 | |
sodium;hexafluorophosphate |
Specifications
>200°C | |
White | |
>99% | |
MFCD00011122 | |
Soluble in water. | |
[Na+].F[P-](F)(F)(F)(F)F | |
167.95 | |
167.95 | |
Hygroscopic | |
2.37 |
21324-39-0 | |
Crystals | |
F6NaP | |
sodium hexafluorophosphate, sodium hexa-fluoro phosphate, sodium hexafluorophosphate v, napf6, sodium hexaflurophosphate, acmc-1cao2, sodium hexafluoro-, e5-phosphanuide, sodium hexafluoro-$l^ 5-phosphanuide, phosphate 1-, hexafluoro-, sodium 1:1 | |
KMADQUOCJBLXRP-UHFFFAOYSA-N | |
sodium;hexafluorophosphate | |
5147921 | |
Odorless | |
White crystalline | |
Sodium hexafluorophosphate |
Safety and Handling
P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
H302+H312+H332-H314
missing translation for 'dotInformation' : Hazard Class: 8, Packing Group: II
missing translation for 'einecsNumber' : 244-333-1
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only