missing translation for 'onlineSavingsMsg'
Learn More
Sodium hyaluronate, 95%, Thermo Scientific Chemicals Product Code.: 10491281

Sodium hyaluronate, 95%, Thermo Scientific Chemicals

Product Code. 10491281
1 g, Glass bottle
missing translation for 'orderingAttributeHoverText'
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
This item is not returnable. View return policy

Product Code. 10491281

Brand: Thermo Scientific Acros 251770010

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 9067-32-7
Molecular Formula (C14H20NO11Na)n
Molecular Weight (g/mol) 417.30
MDL Number MFCD00875848
InChI Key YWIVKILSMZOHHF-QJZPQSOGSA-N
Synonym hyaluronate tetrasaccharide, d0e9sz, 6-3-acetamido-2-6-3-acetamido-2,5-dihydroxy-6-hydroxymethyl oxan-4-yl oxy-2-carboxy-4,5-dihydroxyoxan-3-yl oxy-5-hydroxy-6-hydroxymethyl oxan-4-yl oxy-3,4,5-trihydroxyoxane-2-carboxylic acidShow MoreShow Less
IUPAC Name Sodium hyaluronate
SMILES CC(=O)NC1C(OC2C(O)C(O)C(O-*)OC2C(=O)O[Na])OC(CO)C(O)C1O-*

Specifications

Color White to Cream
pH 6.0 to 8.0 (0.5% aq. soln.)
Loss on Drying 10% max. (500mg, 105°C, 6 hrs)
Infrared Spectrum Authentic
Assay Percent Range 94% min. (On dry substance)
Packaging Glass bottle
Linear Formula (C14H20NO11Na)n
Quantity 1 g
Merck Index 15,4796
Solubility Information Solubility in water: soluble.
Percent Purity 95%
Physical Form Powder
Chemical Name or Material Sodium hyaluronate
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0469414 Certificate TypeCertificate of Analysis Date31/01/2025 Catalog Number251770010,251770050,251770250
Lot NumberA0463288 Certificate TypeCertificate of Analysis Date04/06/2024 Catalog Number251770010,251770050,251770250
Lot NumberA0458406 Certificate TypeCertificate of Analysis Date12/12/2023 Catalog Number251770010,251770050,251770250
Lot NumberA0452930 Certificate TypeCertificate of Analysis Date10/05/2023 Catalog Number251770010,251770050,251770250
Lot NumberA0449530 Certificate TypeCertificate of Analysis Date27/02/2023 Catalog Number251770010,251770050,251770250
Product Identifier
  • Sodium hyaluronate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Sodium hyaluronate, 95%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.