missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Hydroxide Solution 0.5M (0.5N), NIST Standard Concentrate, for Volumetric Analysis, Fisher Chemical™
£34.90 - £189.00
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
10306190
|
Fisher Chemical
J/7630C/05 |
1 Ampule |
£34.90
1 ampoule |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10030200
|
Fisher Chemical
J/7630C/90 |
6 Ampules |
£189.00
6 ampoules |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Chemical Identifiers
3548 |
Specifications
Colorless | |
3548 | |
Ampoule | |
Liquid |
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- Sodium hydroxide solution 0.5M (0.5N) ampoule Solutrate
Signal Word
- Danger
Hazard Category
- Skin corrosion/irritation Category 1 A
- Serious eye damage/eye irritation Category 1
- Substances and mixtures corrosive to metals Category 1
Hazard Statement
- H314-Causes severe skin burns and eye damage.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P315-Get immediate medical advice/attention.
Supplemental information
- MIXTURE LIST-Contains : Sodium hydroxide
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title