missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium hypophosphite monohydrate, Puriss. p.a., Reag. Ph. Eur., ≥99%, Honeywell Fluka™
£27.64 - £2944.80
Chemical Identifiers
CAS | 10039-56-2 |
---|---|
Molecular Formula | H4NaO3P |
Molecular Weight (g/mol) | 105.99 |
MDL Number | MFCD06252379 |
InChI Key | PLZNPHDJGFDNRM-UHFFFAOYSA-M |
Synonym | sodium hypophosphite hydrate, sodium phosphinate hydrate, sodium phosphenite hydrate, sodium hypophosphite en grade, sodium hydrate hypophosphite, sodium hypophosphite monohydrate 250g, phosphinic acid sodium salt monohydrate, sodium hypophosphite monohydrate, phosphonous acid monosodium salt monohydrate, sodium hypophosphite monohydrate, puriss |
PubChem CID | 23708894 |
IUPAC Name | sodium hydrate phosphinate |
SMILES | O.[Na+].[O-][PH2]=O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15603020
|
Honeywell-Fluka
30410-100G |
100 g | N/A | ||||||
15623020
|
Honeywell-Fluka
30410-250G |
250 g | N/A | ||||||
15613020
|
Honeywell-Fluka
30410-1KG |
1 kg | N/A | ||||||
15633020
|
Honeywell-Fluka
30410-50KG |
50 kg |
£2944.80
50kg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Chemical Identifiers
10039-56-2 | |
105.99 | |
PLZNPHDJGFDNRM-UHFFFAOYSA-M | |
23708894 | |
O.[Na+].[O-][PH2]=O |
H4NaO3P | |
MFCD06252379 | |
sodium hypophosphite hydrate, sodium phosphinate hydrate, sodium phosphenite hydrate, sodium hypophosphite en grade, sodium hydrate hypophosphite, sodium hypophosphite monohydrate 250g, phosphinic acid sodium salt monohydrate, sodium hypophosphite monohydrate, phosphonous acid monosodium salt monohydrate, sodium hypophosphite monohydrate, puriss | |
sodium hydrate phosphinate |
Specifications
10039-56-2 | |
NaH2PO2 · H2O | |
NONH for all modes of transport | |
PLZNPHDJGFDNRM-UHFFFAOYSA-M | |
sodium hydrate phosphinate | |
23708894 | |
≥99% | |
Sodium Hypophosphite Monohydrate, Reag. Ph. Eur. |
H4NaO3P | |
MFCD06252379 | |
sodium hypophosphite hydrate, sodium phosphinate hydrate, sodium phosphenite hydrate, sodium hypophosphite en grade, sodium hydrate hypophosphite, sodium hypophosphite monohydrate 250g, phosphinic acid sodium salt monohydrate, sodium hypophosphite monohydrate, phosphonous acid monosodium salt monohydrate, sodium hypophosphite monohydrate, puriss | |
O.[Na+].[O-][PH2]=O | |
105.99 | |
105.99g/mol | |
puriss. p.a. |
Safety and Handling
missing translation for 'einecsNumber' : 231-669-9
missing translation for 'rtecsNumber' : SZ5640000
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Sodium hypophosphite monohydrate, Puriss. p.a., Reag. Ph. Eur., ≥99%, Honeywell Fluka™