Learn More
Sodium iodide, ultra dry, 99.99% (metals basis), Thermo Scientific Chemicals
£99.39 - £982.89
Chemical Identifiers
CAS | 7681-82-5 |
---|---|
Molecular Formula | INa |
Molecular Weight (g/mol) | 149.894 |
MDL Number | MFCD00003532 |
InChI Key | FVAUCKIRQBBSSJ-UHFFFAOYSA-M |
Synonym | sodium iodide, ioduril, sodium monoiodide, soiodin, sodiumiodide, natrii iodidum, jodid sodny, iodure de sodium, jodid sodny czech |
PubChem CID | 5238 |
ChEBI | CHEBI:33167 |
IUPAC Name | sodium;iodide |
SMILES | [Na+].[I-] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11430021
|
Thermo Scientific Alfa Aesar
044766.14 |
25 g |
£99.39
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11440021
|
Thermo Scientific Alfa Aesar
044766.22 |
100 g |
£294.23
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11450021
|
Thermo Scientific Alfa Aesar
044766.36 |
500 g |
£982.89
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Sodium iodide commonly is used to prevent iodine deficiency. Used as analytical reagents, in the manufacture of disinfectants, antiseptics, deodorants and medicines. They are used in baking ingredient conditioner and as animal feed. It is used for conversion of alkyl chlorides into alkyl iodides, such as in the treatment of thyroid cancer and hyperthyroidism and in scintillation detectors, traditionally in nuclear medicine, geophysics, nuclear physics, and environmental measurements.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
7681-82-5 | |
149.894 | |
FVAUCKIRQBBSSJ-UHFFFAOYSA-M | |
5238 | |
sodium;iodide |
INa | |
MFCD00003532 | |
sodium iodide, ioduril, sodium monoiodide, soiodin, sodiumiodide, natrii iodidum, jodid sodny, iodure de sodium, jodid sodny czech | |
CHEBI:33167 | |
[Na+].[I-] |
Specifications
1300°C to 1304°C | |
7681-82-5 | |
Powder | |
INa | |
sodium iodide, ioduril, sodium monoiodide, soiodin, sodiumiodide, natrii iodidum, jodid sodny, iodure de sodium, jodid sodny czech | |
FVAUCKIRQBBSSJ-UHFFFAOYSA-M | |
sodium;iodide | |
5238 | |
149.89 | |
1.7745 | |
3.67 |
651°C | |
White | |
99.99% (metals basis) | |
MFCD00003532 | |
Soluble in water,alcohol,glycerol and acetone. Insoluble in ether. | |
[Na+].[I-] | |
149.894 | |
CHEBI:33167 | |
Odorless | |
Hygroscopic | |
Sodium iodide |
Safety and Handling
In case of contact with eyes, rinse immediately with plenty of water and seek medical advice. Wear suitable gloves.
P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362
H315-H319
missing translation for 'dotInformation' : Hazard Class: 9; Packaging Group: III
missing translation for 'einecsNumber' : 231-679-3
missing translation for 'rtecsNumber' : WB6475000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
- Sodium iodide
Signal Word
- Danger
Hazard Category
- ACUTE AQUATIC Acute 1
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 1
Hazard Statement
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H372-Causes damage to organs.
- H400-Very toxic to aquatic life.
Precautionary Statement
- P264-Wash face, hands and anyexposed skin thoroughly after handling.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P314-Get medical advice/attention if you feel unwell.
Supplemental information
- MIXTURE LIST-Contains : Sodium iodide
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.