missing translation for 'onlineSavingsMsg'
Learn More
Sodium pentacyanonitrosylferrate(III) dihydrate, 98+%, Thermo Scientific Chemicals Product Code.: 11453970

Sodium pentacyanonitrosylferrate(III) dihydrate, 98+%, Thermo Scientific Chemicals

Product Code. 11453970
25 g
Click to view available options
Quantity:
25 g
50 g
100 g
250 g
500 g
This item is not returnable. View return policy

Product Code. 11453970

Brand: Thermo Scientific Alfa Aesar A15656.14

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Sodium pentacyanonitrosylferrate(III) dihydrate, is used as a vasodilator drug to reduce blood pressure. It is used as an analytical reagent for the detection of methyl ketones and amines. Further, it is used as a reference compound for the calibration of Mossbauer spectrometer. It is also used for chromatographic detection of peptides containing cystine. It finds application as a color reagent in the development of thin layer chromatography in combination with acetaldehyde. It is also used in presence of buffers as reagent for ketone strips, which is utilized to test ketone in the urine of a diabetic.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Sodium pentacyanonitrosylferrate(III) dihydrate, is used as a vasodilator drug to reduce blood pressure. It is used as an analytical reagent for the detection of methyl ketones and amines. Further, it is used as a reference compound for the calibration of Mossbauer spectrometer. It is also used for chromatographic detection of peptides containing cystine. It finds application as a color reagent in the development of thin layer chromatography in combination with acetaldehyde. It is also used in presence of buffers as reagent for ketone strips, which is utilized to test ketone in the urine of a diabetic.

Solubility
Soluble in water. Slightly soluble ethanol.

Notes
Hygroscopic and moisture sensitive. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13755-38-9
Molecular Formula C5H4FeN6Na2O3
Molecular Weight (g/mol) 297.95
MDL Number MFCD00149192
InChI Key JFPDVRGGFBUORC-UHFFFAOYSA-N
Synonym nitroprusside sodium, sodium pentacyanonitrosylferrate, sodium nitroferricyanide iii dihydrate, dsstox_cid_21126, dsstox_rid_79630, dsstox_gsid_41126, sodium pentacyanonitrosylferrate iii dihydrate, nitropress tn, sodium nitroprusside usp, c5fen6o.2na.2h2oShow MoreShow Less
PubChem CID 11953895
IUPAC Name disodium;iron(4+);nitroxyl anion;pentacyanide;dihydrate
SMILES O.O.[Na+].[Na+].N#C[Fe++](C#N)(C#N)(C#N)(C#N)[N]#[O+]

Specifications

Density 1.72
Odor Odorless
Linear Formula Na2[Fe(CN)5NO]·2H2O
Quantity 25 g
UN Number UN1588
Sensitivity Hygroscopic
Merck Index 14,8649
Solubility Information Soluble in water. Slightly soluble ethanol.
Formula Weight 297.95 (261.92 Anhydrous)
Percent Purity ≥98%
Chemical Name or Material Sodium pentacyanonitrosylferrate(III) dihydrate
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10253439 Certificate TypeCertificate of Analysis Date23/12/2024 Catalog NumberA15656.22,A15656.14,A15656.36
Lot Number10240721 Certificate TypeCertificate of Analysis Date24/10/2023 Catalog NumberA15656.22,A15656.14,A15656.36
Lot Number10243023 Certificate TypeCertificate of Analysis Date12/09/2022 Catalog NumberA15656.22,A15656.14,A15656.36
Lot Number10240235 Certificate TypeCertificate of Analysis Date27/04/2022 Catalog NumberA15656.22,A15656.14,A15656.36
Lot Number10171572 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberA15656.22,A15656.14,A15656.36

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Sodium pentacyanonitrosylferrate(III) dihydrate, 98+%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.