missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium phosphite dibasic pentahydrate, 98%, Thermo Scientific Chemicals
£20.90 - £52.96
Chemical Identifiers
CAS | 13517-23-2 |
---|---|
Molecular Formula | HNa2O3P·5H2O |
Molecular Weight (g/mol) | 216.02 |
Synonym | sodium phosphite dibasic pentahydrate, di-sodium hydrogen phosphite, di-sodium hydrogen phosphite pentahydrate, disodium pentahydrate hydrogen phosphite, 2na.ho3p.5h2o, di-sodiumhydrogenphosphitepentahydrate, disodium hydrogenphosphite pentahydrate, etidronate disodium related compound a, united states pharmacopeia usp reference standard |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
11994281
|
Thermo Scientific Acros
428502500 |
250 g | Glass bottle |
£20.90
250g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11984281
|
Thermo Scientific Acros
428500010 |
1 kg | Glass bottle |
£52.96
1kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
13517-23-2 | |
HNa2O3P·5H2O | |
216.02 | |
sodium phosphite dibasic pentahydrate, di-sodium hydrogen phosphite, di-sodium hydrogen phosphite pentahydrate, disodium pentahydrate hydrogen phosphite, 2na.ho3p.5h2o, di-sodiumhydrogenphosphitepentahydrate, disodium hydrogenphosphite pentahydrate, etidronate disodium related compound a, united states pharmacopeia usp reference standard |
Specifications
13517-23-2 | |
HNa2O3P·5H2O | |
sodium phosphite dibasic pentahydrate, di-sodium hydrogen phosphite, di-sodium hydrogen phosphite pentahydrate, disodium pentahydrate hydrogen phosphite, 2na.ho3p.5h2o, di-sodiumhydrogenphosphitepentahydrate, disodium hydrogenphosphite pentahydrate, etidronate disodium related compound a, united states pharmacopeia usp reference standard | |
216.02 | |
98% |
98% | |
15,8793 | |
Solubility in water: soluble | |
216.02 | |
Sodium phosphite dibasic pentahydrate |
Safety and Handling
Product Identifier
- Sodium phosphite dibasic pentahydrate
RUO – Research Use Only