missing translation for 'onlineSavingsMsg'
Learn More
Thermo Scientific Chemicals Sodium pyrophosphate, 200mM buffer soln. Product Code.: 15494509

Thermo Scientific Chemicals Sodium pyrophosphate, 200mM buffer soln.

Product Code. 15494509
100 mL
Click to view available options
Quantity:
100 mL
250 mL
This item is not returnable. View return policy

Product Code. 15494509

Brand: Thermo Scientific Chemicals J62052.AE

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Sodium pyrophosphate, 200mM buffer soln. is used in the laboratory as a buffering agent. DSPP has been used as a chelating agent in antimicrobial studies. Tetrabasic pyrophosphate or TSPP (sc-203404; sc-236921) has shown to be useful in the preparation of an EDTA-sodium pyrophosphate extraction buffer for microcystin analysis of soil samples. It is a popular leavening agent found in baking powders.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Sodium pyrophosphate, 200mM buffer soln. is used in the laboratory as a buffering agent. DSPP has been used as a chelating agent in antimicrobial studies. Tetrabasic pyrophosphate or TSPP (sc-203404; sc-236921) has shown to be useful in the preparation of an EDTA-sodium pyrophosphate extraction buffer for microcystin analysis of soil samples. It is a popular leavening agent found in baking powders.

Solubility
Fully miscible in water. Insoluble in alcohol and ammonia.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7758-16-9

Specifications

Concentration 200mM buffer soln.
Quantity 100 mL
Content And Storage Ambient Temperatures
Form Liquid

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberM16L502 Certificate TypeCertificate of Analysis Date18/01/2025 Catalog NumberJ62052.AE,J62052.AK
Lot NumberM16L501 Certificate TypeCertificate of Analysis Date17/01/2025 Catalog NumberJ62052.AE,J62052.AK
Lot NumberW16K544 Certificate TypeCertificate of Analysis Date18/09/2024 Catalog NumberJ62052.AE,J62052.AK
Lot NumberU01K544 Certificate TypeCertificate of Analysis Date05/08/2024 Catalog NumberJ62052.AE,J62052.AK
Lot NumberP02K570 Certificate TypeCertificate of Analysis Date02/04/2024 Catalog NumberJ62052.AE,J62052.AK

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific Chemicals Sodium pyrophosphate, 200mM buffer soln. >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.