missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Pyruvate (100 mM)

Sodium Pyruvate (100 mM)
£9.33 - £156.00
Specifications
Certifications/Compliance | FDA, ISO 13485 |
---|---|
Content And Storage | 2 to 8°C |
Form | Liquid |
Product Type | Sodium Pyruvate |
For Use With (Application) | Mammalian Cell Culture |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11530396
![]() |
Gibco
11360039 |
100 mL |
£9.33
100mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11540396
![]() |
Gibco
11360088 |
20 x 100 mL |
£156.00
Pack of 20 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Sodium Pyruvate is commonly added to cell culture medium as an energy source in addition to glucose. Since the cells make sodium pyruvate as an intermediate metabolite in the glycolysis pathway, it is not necessary to supplement all cultures. However, if the cells have been grown in medium supplemented with sodium pyruvate, we recommend the supplementation is continued as the cells become accustomed to this addition and their growth will lag without it. Gibco™ Sodium Pyruvate, 100 mM Solution is formulated as 11 g per liter of water. The final concentration of sodium pyruvate used in most cell culture medium is 1 mM.Product Use
For in vitro diagnostic use.
Dual-site cGMP Manufacturing and Quality System
Gibco™ Sodium Pyruvate Solution is manufactured at a cGMP compliant facility, located in Paisley, Scotland, UK. The facility is registered with the FDA as a medical device manufacturer and is certified to the ISO 13485 standard. For supply chain continuity, we offer an identical Gibco™ Sodium Pyruvate Solution product made in our Grand Island facility (11360-070). This facility is registered with the FDA as a medical device manufacturer and is certified to the ISO 13485 standards.
Specifications
FDA, ISO 13485 | |
Liquid | |
Mammalian Cell Culture |
2 to 8°C | |
Sodium Pyruvate |
Safety and Handling
missing translation for 'shelfLife' : 12 Months
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- 3,4-Dichlorophenol
Signal Word
- Warning
Hazard Category
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
Hazard Statement
- H302-Harmful if swallowed.
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
- MIXTURE LIST-Contains : 3,4-Dichlorophenol
For In Vitro Diagnostic Use.
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Sodium Pyruvate (100 mM)