missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Sulfate Anhydrous, Certified AR for Analysis, Fisher Chemical™
Click to view available options
Quantity:
500 g
5 kg
Specifications
Specifications
Melting Point | 884°C |
Color | White |
pH | 5 to 8 |
Physical Form | Solid |
Quantity | 5 kg |
IUPAC Name | disodium;sulfate |
Formula Weight | 142.04g/mol |
Packaging | Plastic powder jar |
Product Suggestions
Customers who viewed this item also viewed.
Viewing 1-4 of
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Sodium Sulfate Anhydrous, Certified AR for Analysis, Fisher Chemical™ >
Spot an opportunity for improvement?Share a Content Correction