missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Notrax Sof-Tred™ Microcellular Vinyl Foam with Pebble Pattern, Black/Yellow
Notrax Sof-Tred™ Basic worker comfort derived from 9.4 mm thick worker platform comprised of a construction of microcellular vinyl consisting of millions of air bubbles that provide basic ergonomic benefit.
£27.35 - £61.15
Specifications
Color | Black/Yellow |
---|---|
Material | Microcellular Vinyl |
Type | Industrial and Commercial Matting |
Product Code | Brand | Length (Metric) | Width (Metric) | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Length (Metric) | Width (Metric) | Price | Quantity & Availability | ||||
17722088
|
Notrax
411S0323BY |
91 cm | 60 cm |
£27.35
Each |
Estimated Shipment: 31-07-2024
Log in to see stock. |
||||
17762088
|
Notrax
411S0335BY |
150 cm | 91 cm |
£61.15
Each |
Estimated Shipment: 31-07-2024
Log in to see stock. |
||||
17702078
|
Notrax
411C0324BY |
1830 cm | 60 cm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
17742078
|
Notrax
411C0336BY |
1830 cm | 91 cm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
17782078
|
Notrax
411C0348BY |
1830 cm | 122 cm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
Description
- Pebble pattern provides non-directional traction and is easy to clean
- Basic worker comfort derived from 94 mm thick worker platform comprised of a construction of microcellular vinyl consisting of millions of air bubbles that provide basic ergonomic benefit
- All four sides are bevelled and sealed to minimize tripping hazards
Specifications
Black/Yellow | |
Microcellular Vinyl | |
Industrial and Commercial Matting |