Learn More
Thermo Scientific™ Accessories for Solaris™ 2000 and 4000 Open Air Orbital Shakers
Mix and match a variety of labware on a single platform with flexible Thermo Scientific™ Solaris™ Shaker Universal Platforms and Clamps.
Brand:  Thermo Scientific™ SK3624
 View more versions of this product
Product Code. 16231591
  • £539.07 / 1 unit
Estimated Shipment: 26-08-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
Mix and match a variety of labware on a single platform with flexible Thermo Scientific™ Solaris™ Shaker Universal Platforms and Clamps. With exceptional innovation, reliability and accuracy, these accessories are the optimal choice for a busy lab.
Maximize capacity on the Thermo Scientific™ Solaris Orbital Shakers with universal platforms and a variety of clamps to fit most vessels.
Certifications
UL, cUL, CE
Services
1 Year
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Certificate of Analysis (COA), Certificate of Conformance (COC)
36 in.
24 in.
Solaris 36x24 Universal platform
Platform
36 x 24 in.
914.4 mm
610 mm
Solaris 4000
1 Year
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Thermo Scientific™ Accessories for Solaris™ 2000 and 4000 Open Air Orbital Shakers >