missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Nunc™ Microplate Immuno Maxisorp 96 Well
Brand: Thermo Scientific™ Nunc™ 476635
920.00 GBP valid until 2025-12-31
Use promo code "25339" to get your promotional price.
This item is not returnable.
View return policy
Description
Transferable solid phase (TSP) system facilitates the testing by an ELISA method of two types of analytes simultaneouslyUniform adsorption properties
Ideal for screening of hybridoma cultures for antibody production.

Specifications
Standard | |
Clear | |
96 | |
Round Bottom (U-well) |
8 x 12 | |
Polystyrene | |
MaxiSorp |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-4 of
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Thermo Scientific™ Nunc™ Microplate Immuno Maxisorp 96 Well > U bottom polystyrene transfer for solid phase system (TSP) 21 sleeves of 10 plates clear 95mm2 total surface area Thermo Scientific Nunc
Spot an opportunity for improvement?Share a Content Correction