Learn More
Fisherbrand™ Solutions A+ Aloe Latex Gloves
Protects hands from drying, chapping and cracking. Fisherbrand™ Solutions A+ Aloe Latex Gloves combine protection and comfort.
£29.09
Specifications
Certifications/Compliance ISO 9001 QMS, FDA Biocompatibility Guidance, Passes Viral Penetration Test (ASTM F1671), 510K Approved
Color Natural
Hand Ambidextrous
Material Latex
Powdered Powder Free
View More Specs
Products 4
Product Code Brand Size Price Quantity & Availability  
Product Code Brand Size Price Quantity & Availability  
15163269
View Documents
Fisherbrand™
FB51957NEW
Large This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
15173269
View Documents
Fisherbrand™
FB51955NEW
Medium This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
15153269
View Documents
Fisherbrand™
FB51953NEW
Small This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
15183269
View Documents
Fisherbrand™
FB51959NEW
X-Large This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description
Description
Latex gloves provide a protective barrier and contain soothing Aloe vera to help prevent skin from drying, cracking and chapping. Textured to provide excellent grip. Powder free. EN 374:2003 (AQL 1.5).
  • PPE Category III
  • Disposable
  • Aloe vera coating protects from drying, chapping and cracking
  • Ideal for prolonged use
  • Textured surface provides good grip
  • Latex
Specifications
Specifications
ISO 9001 QMS, FDA Biocompatibility Guidance, Passes Viral Penetration Test (ASTM F1671), 510K Approved
Ambidextrous
Powder Free
5.5 mil
Beaded/Rolled
240 mm
Natural
Latex
Non-sterile
Ideal for workers who have frequent glove changes
18 MPa