missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva SOURCE™ Tricorn™ Columns for IEX
Shop All Cytiva Products
Click to view available options
Product Type:
Source 15Q prepacked column
Source 15S prepacked column
Ligand Type:
Quaternary Ammonium
Sulphonate group
pH Range:
2 to 12
2 to 13
Unit Size:
Each
Description
Includes
Tricorn columns have Valco fittings for AKTAdesign™ systems and are supplied with M6 connectors for FPLC Systems, instruction book, and two stop plugs attached to the column.
- SOURCE 15Q media is highly suitable for fast, high-resolution purifications and easy scale-up
- Rigid, monodispersed, spherical particles with controlled pore-size distribution offer excellent flow characteristics, sample loading of up to 25 mg of protein/ml, and improved stability in organic solvents and pH extremes
- Improved capacity compared to MonoBeads but with a slightly lower resolution.
Recommended Storage
4 to 30°C, 20% Ethanol
Specifications
Specifications
| Buffer | All common ion exchange Buffers |
| Product Type | Source 15Q prepacked column |
| Bed Volume | 1.7mL |
| Diameter (English) Inner | 0.18 in. |
| Diameter (Metric) Inner | 4.6mm |
| Quantity | 1 x 1.7mL |
| Height (English) | 3.93 in. |
| Height (Metric) | 100mm |
| Flow Rate | <5 mL/min. |
| Material | PEEK |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction