missing translation for 'onlineSavingsMsg'
Learn More
Thermo Scientific Chemicals Spironolactone, 99% Product Code.: 10424165

Thermo Scientific Chemicals Spironolactone, 99%

Product Code. 10424165
5 g, Glass bottle
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
This item is not returnable. View return policy

Product Code. 10424165

Brand: Thermo Scientific Chemicals 207460050

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 52-01-7
Molecular Formula C24H32O4S
Molecular Weight (g/mol) 416.57
MDL Number MFCD00082250
InChI Key LXMSZDCAJNLERA-ZHYRCANASA-N
Synonym spironolactone, aldactone, spirolactone, verospiron, euteberol, spiroctan, spirolang, verospirone, aldactone a, spironocompren
PubChem CID 5833
ChEBI CHEBI:9241
IUPAC Name S-[(7R,8R,9S,10R,13S,14S,17R)-10,13-dimethyl-3,5'-dioxospiro[2,6,7,8,9,11,12,14,15,16-decahydro-1H-cyclopenta[a]phenanthrene-17,2'-oxolane]-7-yl] ethanethioateShow MoreShow Less
SMILES CC(=O)SC1CC2=CC(=O)CCC2(C3C1C4CCC5(C4(CC3)C)CCC(=O)O5)C

Specifications

Melting Point 198.0°C to 207.0°C
Color White to White-Yellow
Loss on Drying 0.5% max. (105°C, 2 hrs)
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (HPLC)
Packaging Glass bottle
Quantity 5 g
Merck Index 15, 8887
Specific Rotation Condition −37° (20°C c=1,CHCl3)
Specific Rotation − 37.00
Solubility Information Solubility in water: practically insoluble. Other solubilities: soluble in most common organic solvents
Formula Weight 416.57
Percent Purity 99%
Physical Form Powder
Chemical Name or Material Spironolactone, 99%
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0462192 Certificate TypeCertificate of Analysis Date29/04/2024 Catalog Number207460010,207460050
Lot NumberA0433068 Certificate TypeCertificate of Analysis Date15/02/2024 Catalog Number207460010,207460050
Lot NumberA0401224 Certificate TypeCertificate of Analysis Date14/12/2022 Catalog Number207460010,207460050
Lot NumberA0409558 Certificate TypeCertificate of Analysis Date13/12/2022 Catalog Number207460010,207460050
Lot NumberA0430587 Certificate TypeCertificate of Analysis Date08/06/2021 Catalog Number207460010,207460050
compliance-icons compliance-icons
Product Identifier
  • Spironolactone
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Reproductive toxicity Category 1B
Hazard Statement
  • H360F-May damage fertility.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific Chemicals Spironolactone, 99% >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.