Learn More
BRAND™ Spotting Tile
12 polished cavities
Brand:  BRAND™ 473500
Product Code. 10223731
  • £109.00 / Each
In Stock
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Soda-lime glass
  • 12 polished cavities 20 - 22 mm diameter, depth 2 mm, capacity approx. 0.2 mL
  • Ground rectangular edges
  • TRUSTED_SUSTAINABILITY
    Specifications
    Specifications
    Spotting Tile
    Soda Lime Glass
    1
    130 x 100 x 6 mm
    Product Suggestions
    Product Suggestions
    Videos
    SDS
    Documents
    Documents
    Certificates
    Special Offers
    Special Offers
    Product Content Correction
    Product Title
    BRAND™ Spotting Tile > Dimensions (L x W x H) Exterior: 130 x 100 x 6 mm