Learn More
Brady™ Universal Plus Chemical Roll
The Universal Plus Chemical Roll is a 2-ply, green, medium weight, bonded and double perforated chemical sorbent roll that absorbs chemicals and hazardous liquids.
Brand: Brady™ 813845
Description
- One sided spunbound gives extra strength as well as less lint.
- Does not degrade or cause a dangerous chemical reaction with the absorbed liquid.
- Green color alerts workers that the product is being used for hazardous liquids.
- Green color also allows the user to identify and separate hazardous waste for disposal, resulting in lower disposal costs.
- Perforated down the middle and cross-wise to permit the use of precisely the right amount of material to reduce waste.
- Perfect for cleaning up chemical spills or for use in laboratories.
Specifications
UN15-DP-E | |
ROLL, Medium weight, double perforated and bonded 38 cm x 46 m | |
78 L | |
Absorbent Roll | |
Overspray, leaks, drips, in drum and liquid storage areas, workstations and laboratories. | |
46 m x 38 cm | |
46 m | |
38 cm |
Green | |
20.605 gal. | |
Chemical Sorbent | |
Beverage processing, Chemical, Food processing, Laboratory, Manufacturing, Maritime, Oil & gas, Pharmaceutical, Process, Warehouse & logistics | |
1 Roll | |
50.31 yd. | |
14.96 in. | |
6.3 kg |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.