missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Stained Salmonella O and H Suspensions

Rapidly detect and quantify specific antibodies to Salmonella in sera with this standardized stained suspension.

Brand:  Thermo Scientific™ R30855301

 View more versions of this product

Product Code. 11993262

  • £135.00 / 5mL

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Rapidly detect and quantify specific antibodies to Salmonella spp. in sera with Thermo Scientific™ Remel™ Stained Salmonella O and H Suspensions. Stained Salmonella O and H Suspensions are standardized, smooth suspensions of killed bacteria which have been stained to facilitate reading of agglutination tests.

Use Stained Salmonella O and H Suspensions for the qualitative detection of Salmonella antibodies in sera for epidemiological and diagnostic purposes, primarily in the investigation of enteric infections and pyrexia.

  • Identifies major groups of Salmonella serotypes
  • Provides accurate, reliable results
  • Simple, easy-to-use tests

Serological tests are based on the fact that antibodies in serum, produced in response to exposure to bacterial antigens, will agglutinate bacterial suspensions which carry homologous antigens. In the diagnosis of febrile disease such tests should be used in parallel with appropriate cultural techniques for the isolation and identification of the causative organism. The suspensions are suitable for use in rapid slide tests1 and standard Widal tests2, 3. They may also be used as control antigens for serological identification of bacterial isolates.

 Not all products are available for sale in all territories. Please inquire.

Remel™ and Oxoid™ products are now part of the Thermo Scientific™ brand.

TRUSTED_SUSTAINABILITY
Specifications

Specifications

2°C to 8°C
S. typhi-O
5 mL
Stained Salmonella O and H Suspensions
Stained Salmonella O and H Suspensions, instruction for use
Salmonella
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Instructions

Certificates

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number3310388 Certificate TypeCertificate of Analysis Date20/05/2022 Catalog NumberR30855301
Lot Number3209264 Certificate TypeCertificate of Analysis Date22/12/2020 Catalog NumberR30855301
Lot Number2915976 Certificate TypeCertificate of Analysis Date07/01/2020 Catalog NumberR30855301
Lot Number2902121 Certificate TypeCertificate of Analysis Date18/12/2019 Catalog NumberR30855301
Lot Number2868929 Certificate TypeCertificate of Analysis Date31/10/2019 Catalog NumberR30855301
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific™ Stained Salmonella O and H Suspensions >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.