Learn More
Hamilton™ Stainless Steel Luer Lock Metal Hub Needle, Custom length, Point Style 2, 3, or 4
Metal hub makes the most rigid connection to the needle making it perfect for repetitive septum piercing.
Brand: Hamilton™ 7749-03
This item is not returnable.
View return policy
Description
- Available in various gauges
- Autoclavable
- Metal hubs manufactured from nickel plated brass
- Custom length
- Compatible with PTFE Luer Lock (TLL) syringes
- Design controlled by a series of universal standards which guarantees compatibility between manufacturers
- Provided in packs of 6
Specifications
Autoclavable | |
12 G | |
2.77 mm | |
9.52 to 304 mm | |
TLL Syringes and LT or TLL Connectors |
Stainless Steel | |
2.16 mm | |
Luer Lock Custom Needle | |
2, 3, or 4 | |
Standard Tapered |
Product Suggestions
Customers who viewed this item also viewed.
Viewing 1-4 of
Product Content Correction
Product Title
Hamilton™ Stainless Steel Luer Lock Metal Hub Needle, Custom length, Point Style 2, 3, or 4 > Gauge: 12
Spot an opportunity for improvement?Share a Content Correction