missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Buerkle™ Stainless Steel Safety Cans
For safe metering and handling of smaller amounts of flammable liquids.Protection from exploding through flame protection and excess pressure valve, containers have a high degree of chemical resistance.
£479.00 - £617.00
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | ||||||
11921551
|
Buerkle™
2601-1001 |
1 L |
£479.00
Each |
Estimated Shipment: 13-05-2025
Log in to see stock. |
||||||
15649293
|
Buerkle™
2601-1005 |
5 L |
£617.00
Each |
Estimated Shipment: 02-07-2025
Log in to see stock. |
||||||
11931551
|
Buerkle™
2601-1002 |
2 L |
£518.00
Each |
Estimated Shipment: 16-05-2025
Log in to see stock. |
||||||
Description
- Stainless steel AISI 316 Ti (1.4571), 0.6 mm thickness
- Seals made of PTFE
- Handle made of powder-surfaced Aluminum
- Closure thread 1¼in.
Specifications
Stainless Steel |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Name des Produkts
Buerkle™ Stainless Steel Safety Cans