missing translation for 'onlineSavingsMsg'
Learn More

Tenak™ Stainless Steel Upright Freezer Rack for Cryogenic Boxes

Use Tenak™ Stainless Steel Upright Freezer Racks for Cryogenic Boxes for your cryogenic storage needs. A variety of stainless steel racks accommodate box heights and quantities for laboratory freezers.

Brand:  Tenak™ TE23124

 View more versions of this product

Product Code. 15268373

  • £89.95 / Each
Estimated Shipment: 11-08-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Suitable for all brands of upright freezers
  • Stainless steel
  • For cryogenic boxes of various capacities
Specifications

Specifications

Yes
5 x 3
Stainless Steel
15
Upright Freezers
1 Ea.
276 mm
Standard Rack
Boxes (15)
15
Fit into any freezer brand and model
1
139 mm
419 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Tenak™ Stainless Steel Upright Freezer Rack for Cryogenic Boxes > Holds: Boxes (15); Array: 5 x 3

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.