Learn More
Vici Valco Instruments™ Stainless Steel Tubing Cutter
Achieve smooth and uniform cutting of stainless steel tubing with this cutting wheel type tubing cutter. Vici Valco Instruments™ Stainless Steel Tubing Cutter is ideal for cutting 1/16 in. and 1/8 in. SS tubing.
Brand:  Vici Valco Instruments™ JR-792
Encompass_Preferred
Product Code. 12772680
  • £77.12 / Each
Estimated Shipment: 06-05-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • For 1/16 in. and 1/8 in. SS tubing
  • Leaves smooth, uniform cuts
  • Minimal deburring and/or reaming required
  • Easy replacement of the cutting wheel
  • Creates cuts for tube-to-tube and tube-to-frit connections
  • Creates suitable cuts for zerod dead volume connections
Specifications
Specifications
Tubing Cutter
1/16 in. and 1/8 in. SS Tubing
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Vici Valco Instruments™ Stainless Steel Tubing Cutter