missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Pierce™ Anti-HA Magnetic Beads

Perform immunoprecipitation of recombinant HA-tagged proteins expressed in bacterial or mammalian cells or in vitro systems, using manual or robotic magnetic separators.

Brand:  Thermo Scientific™ 88836

 View more versions of this product

Product Code. 13464229

  • £247.00 / 1mL

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Thermo Scientific Pierce Anti-HA Magnetic Beads are affinity particles for immunoprecipitation of recombinant HA-tagged proteins expressed in bacterial or mammalian cells or in vitro systems, using manual or robotic magnetic separators.

Features of Anti-HA Magnetic Beads:

  • Specific—highly specific anti-HA monoclonal antibody (clone 2-2.2.14) enables high yield and high purity immunoprecipitation
  • Convenient and fast—product instructions provide an easy-to-follow, optimized protocol for immunoprecipitation in approximately one hour
  • Low non-specific binding—stable, pre-blocked beads and specific antibody minimize off-target binding for HA-tag IP or co-IP experiments
  • Versatile—beads are compatible with manual and automated workflows (e.g., Thermo Scientific KingFisher Instruments)

The blocked magnetic bead surface is coated with anti-HA antibody, a highly specific mouse IgG1 monoclonal antibody that recognizes the HA-epitope tag (YPYDVPDYA) derived from the human influenza hemagglutinin (HA) protein. The Pierce Anti-HA Magnetic Beads can be used manually with a magnetic stand, as well as with automated platforms, such as the Thermo Scientific KingFisher Instruments, for high-throughput workflows.

Specifications:

Product Details:

Pierce Anti-HA Magnetic Beads are convenient for the immunoprecipitation (IP) of recombinant HA-tagged proteins and the co-immunoprecipitation (co-IP) of their interacting proteins. The beads are incubated with a cell lysate containing HA-tagged protein and the fusion protein is captured. The beads are subsequently washed and then the target proteins are eluted using 0.1M glycine, pH 2.0, 50 mM NaOH, or SDS-PAGE sample buffer. If gentler elution conditions are desired, 2 mg/mL Pierce HA peptide can be used. The protocol has been optimized for each of these conditions. Anti-HA antibody can be used to detect HA-tagged protein by Western blot analysis.

TRUSTED_SUSTAINABILITY
Specifications

Specifications

1 mL
Store at 4°C.
Liquid
1 mL
HA
Antibody
Slurry: 10mg/mL, 1% solids
1 μm
Pierce™
Recombinant
Magnetic Affinity Bead
Product Suggestions

Product Suggestions

Videos
SDS
Documents Certificates

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberZJ401470 Certificate TypeCertificate of Analysis Date16/12/2024 Catalog Number88836
Lot NumberZI398152 Certificate TypeCertificate of Analysis Date15/10/2024 Catalog Number88836
Lot NumberZH394438 Certificate TypeCertificate of Analysis Date27/08/2024 Catalog Number88836
Lot NumberZC388608 Certificate TypeCertificate of Analysis Date20/03/2024 Catalog Number88836
Lot NumberZA386271 Certificate TypeCertificate of Analysis Date01/02/2024 Catalog Number88836
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific™ Pierce™ Anti-HA Magnetic Beads > 1 mL

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.