Learn More
Integra Biosciences™ Stand
Brand: Integra Biosciences™ 155501
This item is not returnable.
View return policy
Description
VACUBOY hand operator set comprises of: VACUBOY hand operator, stand, rubber adapter for Pasteur pipettes, 1 channel stainless steel aspiration adapter 40mm, 8 channel stainless steel aspiration adapter 40mm, 1 channel plastic aspiration adapter for disposable tips, 1 channel plastic aspiration adapter for disposable tips with ejector, 8 channel plastic aspiration adapter for disposable tips with ejector. Can be used with VACUSAFE comfort vacuum system. Full range of accessories available. Enables liquid residues to be disposed of reliably, precisely and conveniently:- With a precisely dosed volume
- Without loss of drops
- Without risk of contamination
- Without causing tiredness
- Energy saving, the manual operator - the heart of the system - can be operated as a single or 8 channel aspirator
- All parts in contact with fluids are autoclavable
Specifications
Vacuboy bench stand |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-5 of 12
Product Content Correction
Product Title
Integra Biosciences™ Stand > For use with: Parking vacoboy hand operator
Spot an opportunity for improvement?Share a Content Correction