missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Standard Plate Count Agar (APHA) (Dehydrated)

Determine the plate counts of microorganisms in food, dairy samples, waste water and other materials of sanitary importance with this medium.

Brand:  Thermo Scientific™ CM0463B

129.84 GBP valid until 2025-12-31
Use promo code "25339" to get your promotional price.


 View more versions of this product

Product Code. 10679554

  • £279.00 / 500g

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Determine the plate counts of microorganisms in food, dairy samples, waste water and other materials of sanitary importance with this medium.

  • Meets formulation of APHA, AOAC and PHLS
  • Quality control of Standard Plate Count Agar (APHA) includes testing in accordance with ISO 11133:2014
TRUSTED_SUSTAINABILITY
Specifications

Specifications

APHA, AOAC, PHLS, ISO 11133:2014
Agar
500 g
Powder
Standard Plate Count for Microorganism
For 21.3L medium
Product Suggestions

Product Suggestions

Videos
SDS
Documents Certificates

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number6224153 Certificate TypeCertificate of Analysis Date03/03/2025 Catalog NumberCM0463B
Lot Number6195050 Certificate TypeCertificate of Analysis Date29/11/2024 Catalog NumberCM0463B
Lot Number6195049 Certificate TypeCertificate of Analysis Date29/11/2024 Catalog NumberCM0463B
Lot Number6174056 Certificate TypeCertificate of Analysis Date17/10/2024 Catalog NumberCM0463B
Lot Number6157681 Certificate TypeCertificate of Analysis Date23/09/2024 Catalog NumberCM0463B
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific™ Standard Plate Count Agar (APHA) (Dehydrated) > 500 g

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.