missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Implen™ NanoPhotometer™ Standard Solutions
Use to control the photometric accuracy of the NanoPhotometer™ NP120, NP80, N60 and N50 (NanoVolume)
Brand: Implen™ N-120-S
This item is not returnable.
View return policy
Description
- Standard solutions for the control of photometric accuracy
- Verifies the instrument's absorbance accuracy
- For use with NanoPhotometer™ NP120/NP80/N60/N50
- CAS-No: 877-24-7
Specifications
NIST Traceable | |
NanoPhotometer N120 | |
5 x 1 mL (Per Solution) | |
280 nm |
Non-toxic Potassium Hydrogen Phthalate Solution (PHP) | |
Standard Solutions I and II | |
Store at room temperature |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Implen™ NanoPhotometer™ Standard Solutions > NanoPhotometer N120, 5 x 1 mL (Per Solution)
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction