Learn More
3M™ Petrifilm™ Staph Express Count Plate
Provide Staphylococcus aureus results. S. aureus appears as distinctive red-violet colonies. Other colonies have to be checked using the Staph express disk.
Brand:  3M™ Petrifilm™ 7100090291
Encompass_Preferred
 View more versions of this product
Product Code. 17253774
  • £108.17 / Pack of 50
Explore more special offers
Description
Description
  • Results in 24 Hours.
  • Plates are sample-ready, eliminating all the time-consuming steps of preparing media/agar dishes.
  • Unique system to facilitate the enumeration of S. aureus
  • This plate has been awarded Official Methods of Analysis (OMA) #2003.07 for processed and prepared foods, #2003.8 for select dairy foods and #2003.11 for Select meat, seafood , poultry and NF VALIDATION certificates: 3M-01/9-04/03 A and 3M-01/9-04/03 B for All human foods and Pet food by AFNOR Certification.
  • Food processors and contract labs worldwide trust 3M Petrifilm Plates to provide accurate, efficient, consistent results.
Specifications
Specifications
Food and Beverage, Environmental Test
50
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
3M™ Petrifilm™ Staph Express Count Plate