Learn More
3M™ Petrifilm™ Staph Express Disk
Used to identify Staphylococcus aureus from all suspect colonies for samples with high background flora.
Brand:  3M™ Petrifilm™ 7100039573
Encompass
 View more versions of this product
Product Code. 17203764
  • POA / Pack of 100
This item requires a price update prior to being ordered.
Request a Price for this item here.
View alternative products »
Explore more special offers
Description
Description
  • Utilizes the convenience and efficiency of the 3M™ Petrifilm™ Plate platform.
  • Proven cost effective tool for the detection of Staph. aureus.
  • Results in as few as 22 hours.
Specifications
Specifications
Pet Food
100
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
3M™ Petrifilm™ Staph Express Disk