Learn More
Contec™ Sterile IPA

Provided sterile by gamma irradiated using a validated process at no less than 25 kGy. Contec™ Sterile IPA is an alcohol blend of 0.2 micron filtered, filled and bagged in a Grade C (ISO Class 7) cleanroom.
Brand: Contec™ SBC570I
168.75 GBP valid until 2025-05-02
Use promo code "24151" to get your promotional price.
Description
- Guaranteed endotoxin levels of <0.25EU/mL make it ideal for use in product contact areas
- GMP manufactured under Grade A laminar flow air flow in a Grade C cleanroom ensures the alcohol, container and packaging are free from contamination and particulates
- Trigger spray and bag in bottle protected system prevents bottle contents from becoming contaminated during use

Specifications
Alcohol | |
Not Applicable | |
Colorless | |
Clear | |
70% (v/v) IPA and 30% Water | |
Limited-use | |
Yes | |
2 x 5 L | |
5 L |
Liquid | |
Sterile | |
Not Autoclavable | |
Isopropanol in purified water | |
Capped Bottle | |
Filtered | |
Sterilizes surfaces | |
1.3 gal. |
Product Suggestions
Customers who viewed this item also viewed
Safety and Handling
- CONTEC STERILE 70% IPA 1L AND 5L
Signal Word
- Danger
Hazard Category
- Flammable liquid Category 2
- Serious eye damage/eye irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H225-Highly flammable liquid and vapour.
- H319-Causes serious eye irritation.
- H336-May cause drowsiness or dizziness.
- H335-May cause respiratory irritation.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P243-Take precautionary measures against static discharge.
- P370+P378-In case of fire: Use to extinguish.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P403+P233-Store in a well-ventilated place. Keep container tightly closed.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P337+P313-If eye irritation persists: Get medical advice/attention.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
241102927 | Certificate of Quality | 01/03/2025 | SBC570I |
240602771 | Certificate of Quality | 17/10/2024 | SBC570I |
240402735 | Certificate of Quality | 10/08/2024 | SBC570I |
230802545 | Certificate of Quality | 25/05/2024 | SBC570I |
230802536 | Certificate of Quality | 21/03/2024 | SBC570I |
Special Offers
-
Multiple Manufacturers
Cleanroom Essentials Extravaganza
Gear Up Your Controlled EnvironmentPromo Code: 24151Expires: 2025-05-02
Days
32Hours
0Minutes
29Seconds
22New Offer Save Up to 92%
Your input is important to us. Please complete this form to provide feedback related to the content on this product.