missing translation for 'onlineSavingsMsg'
Learn More

Buerkle™ SteriPlast™ Blue Food Scoops
GREENER_CHOICEBIOPHARMA

For sampling powders, granulates and pastes. Buerkle™ SteriPlast™ Food Scoops are sterilized by gamma rays. The color blue does not occur in a natural form in foodstuffs. Scoops can be quickly found and easily identified in a visual check if ever mixed into production.

Brand:  Buerkle™ 5378-3007

 View more versions of this product

Product Code. 15674124

  • £22.60 / Pack of 10
In Stock
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Use of blue tools helps to meet requirements of HACCP hazard analysis and assessment in reference to foreign objects
  • This is important criterion in food processing and production, and in animal feed and pharmaceutical industry
  • Can be used as part of HACCP/IFS foreign object management
  • Individually packaged for disposable use
  • Production, assembly and packaging according to clean room class 7
  • Can be detected by colour scanners

Compliance

EU food requirements

Specifications

Specifications

Food Sampling Scoop
Not Autoclavable
150 mL
Sterile
Sampling Powders, Granulates and Pastes
216 mm
Polystyrene
5.07 oz.
Single-use
Blue
8.5 in.
Product Suggestions

Product Suggestions

Videos

Videos

  • Buerkle SteriPlast Detectable Scoops

SDS
Documents Certificates

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number241705 Certificate TypeCertificate of Compliance Date04/07/2024 Catalog Number5378-3007
Lot Number234339 Certificate TypeCertificate of Compliance Date27/10/2023 Catalog Number5378-3007
Lot Number230342 Certificate TypeCertificate of Compliance Date15/02/2023 Catalog Number5378-3007
Lot Number221007 Certificate TypeCertificate of Compliance Date02/05/2022 Catalog Number5378-3007
Lot Number224356 Certificate TypeDeclaration of Conformity Date07/12/2022 Catalog Number5378-3007
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Buerkle™ SteriPlast™ Blue Food Scoops > Length: 216mm (8.5 in.); 150mL

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.