Learn More
Sartorius Sterisart™ Easy Configuration Software
Sterisart™ Easy Configuration Software is drag and drop software designed to increase process reliability for the sterility testing of sterile pharmaceuticals.
Brand:  Sartorius 1Z----0004
Product Code. 17288720
  • £4937.14 / Each
Explore more special offers
Description
Description
  • Step-by-step guidance through each sterility test programmed, such as for USP 71 or Eu. Ph. 2.6.1
  • FDA 21 CFR Part 11 compliant electronic records
  • User account management, audit trail and password protection
  • Scanning, saving and printing of process-relevant data and records on materials
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Automate Online Configuration Tool
Sterisart Universal Pump
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Sartorius Sterisart™ Easy Configuration Software > Sterisart™ Easy Configuration Software