missing translation for 'onlineSavingsMsg'
Learn More

Sampling Systems™ SteriWare™ Forceps (Tweezers)
BIOPHARMA

Our Sterile single use SteriWare™ Forceps (Tweezers) are completely manufactured in a cleanroom to ensure cleanliness. They come with full documentation that confirms food grade and sterility.

£58.40 - £71.35

Specifications

Material Polystyrene
Length (Metric) 115 mm
Packaging Type Bag
View More Specs

Products 2
Product Code Brand Sterility Price Quantity & Availability  
Product Code Brand Sterility Price Quantity & Availability  
17594676
BIOPHARMA
View Documents
Sampling Systems™
8630A-115S
Non-sterile
£71.35
Pack of 100
Estimated Shipment: 30-04-2025
to see stock.
Add to basket
 
17584676
BIOPHARMA
View Documents
Sampling Systems™
8630A-115
Sterile
£58.40
Pack of 100
Estimated Shipment: 30-04-2025
to see stock.
Add to basket
 
Description

Description

  • Our disposable forceps are available in both sterile and non sterile versions.
  • They are suitable for lifting and moving small objects such as tablets, samples etc.
  • The handles have undulations moulded into them to give you extra grip.
  • These lightweight tweezers are suitable for food, chemical, pharmaceutical, or cosmetic applications.
  • These tweezers can be used straight from the bag.
  • The sterile tweezers have been processed by gamma irradiation.
  • Made from food grade PS.
  • Conforms to the latest FDA and EU requirements.
  • BSE/TSE free
  • To avoid contamination, they have been manufactured and then packed in a class 7 cleanroom.
  • They come with full documentation that confirms food grade and sterility.
Specifications

Specifications

Polystyrene
115 mm
Bag
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Sampling Systems™ SteriWare™ Forceps (Tweezers)

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.