missing translation for 'onlineSavingsMsg'
Learn More

IBA Lifesciences Strep-Tactin™ TACS Agarose Column

Gravity flow column suitable for traceless affinity cell and exosome isolation

£41.05 - £87.35

Specifications

Formulation Pre-packed in buffer, pH 8: 100 mM Tris-HCl pH 8.0, 1 mM EDTA, 150 mM NaCl, 0.01% sodium azide
Support Type Cross-linked agarose
For Use With (Application) Affinity Chromatographic Cell or Exosome Isolation
View More Specs

Products 2
Product Code Brand Capacity (Metric) Price Quantity & Availability  
Product Code Brand Capacity (Metric) Price Quantity & Availability  
17227750
View Documents
IBA Lifesciences
6-6310-001
1 mL
£87.35
Each
Estimated Shipment: 23-12-2024
to see stock.
Add to basket
 
17287590
View Documents
IBA Lifesciences
6-6310-300
0.3 mL
£41.05
Each
Estimated Shipment: 23-12-2024
to see stock.
Add to basket
 
Description

Description

  • Strep-Tactin™ TACS Agarose columns contain agarose macrobeads coated with the streptavidin variant Strep-Tactin™.
  • A special crosslinking permits the use for cell isolation and depletion approaches.
  • They were developed for column-based Traceless Affinty Cell Selection (Fab-TACS™/Nano-TACS™).
  • In this approach, Fab fragments fused to a Twin-Strep-tag™ (Fab-Streps) or nanobodies (Nano-Streps) are bound to Strep-Tactin™ on the agarose matrix.
  • After specifically capturing cells, Fab-/Nano-Streps are released from the Strep-Tactin™ Agarose by biotin addition.
  • This way the cells are eluted ready for further downstream applications.
  • The same principle is applicable for exosome isolation (Fab-TACS™ for exosomes).

  • Alternatively to Fab-Streps/Nano-Streps, it is possible to load columns with Twin-Strep-tagged, desthiobiotinylated or biotinylated antibodies (irreversible binding).
  • This makes Strep-Tactin™ TACS Agarose columns also suitable for negative cell selection or depletion approaches.
Specifications

Specifications

Pre-packed in buffer, pH 8: 100 mM Tris-HCl pH 8.0, 1 mM EDTA, 150 mM NaCl, 0.01% sodium azide
Cross-linked agarose
Affinity Chromatographic Cell or Exosome Isolation
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only.