missing translation for 'onlineSavingsMsg'
Learn More

IBA Lifesciences Strep-Tactin™XT Conjugate
GREENER_CHOICE

Strep-Tactin™XT conjugates with fluorescent label to detect Strep-tag™II and Twin-Strep-tag™ fusion proteins

Brand:  IBA Lifesciences 2-1568-050

 View more versions of this product

Product Code. 16461475

  • £331.00 / 50µg
Estimated Shipment: 21-01-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Strep-Tactin™XT is a streptavidin variant, which binds to the Strep-tag™II and Twin-Strep-tag™ with high affinity and specificity.

It is available as conjugate with fluorescent dyes (DY-488, DY-549, DY-649) for fast and directed detection of N-terminally, C-terminally as well as internally tagged Strep-tag™II or Twin-Strep-tag™ fusion proteins in applications like ELISA, FACS, immunohistochemistry, and immunocytochemistry.

The absorption/emission maxima of the fluorescent dyes are:
• DY-488: 493/518 nm
• DY-549: 550-560/575 nm
• DY-649: 655/676 nm

Specifications

Specifications

0.10 mL
Strep-Tactin™XT conjugated with DY-649
Undiluted: 2°C to 8°C, protect from light; Diluted: -25°C to -15°C
ELISA, western blot, FACS, immunofluorescence
Conjugate
0.5 mg/mL
Solution in PBS; 0.01 % NaN3; 1 mg/ mL BSA
50 μg
Strep-tag™II and Twin-Strep-tag™
DY-649
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only.