Learn More
Thermo Scientific Chemicals Streptomycin sulfate
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
General Description
• Streptomycin sulfate is an aminoglycoside sulfate salt derived from Streptomyces griseus that shows antibacterial activity.
• It inhibits initiation and causes misreading of rRNA in protein synthesis. This compound binds to the S12 protein of the bacterial 30S ribosomal subunit, thereby inhibiting peptide elongation and protein synthesis.
Application
• Streptomycin sulfate can be used to study the mechanisms of streptomycin resistance
• This compound can be used in laboratory applications combined with penicillin to inhibit bacterial contamination in in vitro cell culture
• Streptomycin sulfate shows antibiotic activity against Mycobacterium tuberculosis
• This compound can be used as a protein synthesis inhibitor
• It acts against gram-negative and gram-positive bacteria • Streptomycin sulfate also shows a fungicide activity
• It can be used to remove nucleic acids from a biological extract

Specifications
Specifications
Type | Streptomycin Sulfate |
Loss on Drying | 7% max. |
Quantity | 5 g |
Solubility Information | Solubility in water: soluble. |
Formula Weight | 1457.4 |
Chemical Name or Material | Streptomycin sulfate |
Product Suggestions
Customers who viewed this item also viewed.
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
A0462621 | Certificate of Analysis | 17/05/2024 | 455340050,455340250,455341000 |
A0458703 | Certificate of Analysis | 18/12/2023 | 455340050,455340250,455341000 |
A0455487 | Certificate of Analysis | 25/08/2023 | 455340050,455340250,455341000 |
A0452192 | Certificate of Analysis | 02/05/2023 | 455340050,455340250,455341000 |
A0450191 | Certificate of Analysis | 02/05/2023 | 455340050,455340250,455341000 |
Safety and Handling


- Streptomycin Sulfate
- Warning
- Acute toxicity Category 4
- Reproductive toxicity Category 2
- Respiratory sensitiser Category 1
- H317-May cause an allergic skin reaction.
- H361d-Suspected of damaging the unborn child.
- H302-Harmful if swallowed.
- P201-Obtain special instructions before use.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
Special Offers
-
Multiple Manufacturers
Chromatography and Chemicals Promotion
Explore our new promotion and get up to 70% off some of our most popular products.Promo Code: 24127Expires: 2025-05-31
New Offer Save Up to 70%
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.