missing translation for 'onlineSavingsMsg'
Learn More
Styrene, 99%, stab. with 10-15ppm 4-tert-butylcatechol, Thermo Scientific Chemicals Product Code.: 11408397

Styrene, 99%, stab. with 10-15ppm 4-tert-butylcatechol, Thermo Scientific Chemicals

Product Code. 11408397
500 mL
Click to view available options
Quantity:
100 mL
500 mL
2500 mL
This item is not returnable. View return policy

Product Code. 11408397

Brand: Thermo Scientific Alfa Aesar A18481.AP

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Styrene is used in the preparation of polystyrene which finds application in rigid foodservice containers, automobile body panels, wind turbine parts, boats, compact disc (CD) cases, envelope windows and many other products. Other uses include refrigerator liners, medical devices and small household appliances. It is used as a precursor for the preparation of styrene-butadiene rubber, styrene-butadiene latex, styrene-isoprene-styrene (SIS), styrene-divinylbenzene (S-DVB) and styrene-acrylonitrile resin (SAN), which are used in rubber, plastic, insulation, fiberglass, pipes, automobile and boat parts, food containers and carpet backing.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Styrene is used in the preparation of polystyrene which finds application in rigid foodservice containers, automobile body panels, wind turbine parts, boats, compact disc (CD) cases, envelope windows and many other products. Other uses include refrigerator liners, medical devices and small household appliances. It is used as a precursor for the preparation of styrene-butadiene rubber, styrene-butadiene latex, styrene-isoprene-styrene (SIS), styrene-divinylbenzene (S-DVB) and styrene-acrylonitrile resin (SAN), which are used in rubber, plastic, insulation, fiberglass, pipes, automobile and boat parts, food containers and carpet backing.

Solubility
Miscible with carbon disulfide, ether, methanol, benzene, toluene, ethanol, n-heptane, carbon tetrachloride and acetone. Immiscible with water.

Notes
Air and light sensitive. Store in cool place. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with oxidizing agents and copper.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 100-42-5
Molecular Formula C8H8
Molecular Weight (g/mol) 104.15
MDL Number MFCD00008612,MFCD00084450
InChI Key PPBRXRYQALVLMV-UHFFFAOYSA-N
Synonym ethenylbenzene, phenylethylene, vinylbenzene, styrol, benzene, ethenyl, cinnamene, phenylethene, monomer, phenethylene, styroleneShow MoreShow Less
PubChem CID 7501
ChEBI CHEBI:27452
SMILES C=CC1=CC=CC=C1

Specifications

Melting Point -31°C
Density 0.906
pH Neutral
Boiling Point 145°C to 146°C
Flash Point 31°C (88°F)
Odor Pungent
Refractive Index 1.546
Quantity 500 mL
UN Number UN2055
Beilstein 1071236
Sensitivity Light sensitive
Merck Index 14,8860
Solubility Information Miscible with carbon disulfide,ether,methanol,benzene,toluene,ethanol,n-heptane,carbon tetrachloride and acetone. Immiscible with water.
IUPAC Name styrene
Formula Weight 104.15
Percent Purity 99%
Chemical Name or Material Styrene, stabilized with 10 to 15ppm 4-tert-butylcatechol
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10234723 Certificate TypeCertificate of Analysis Date30/08/2024 Catalog NumberA18481.0F,A18481.AE,A18481.AP
Lot Number10245503 Certificate TypeCertificate of Analysis Date13/11/2023 Catalog NumberA18481.0F,A18481.AE,A18481.AP
Lot Number10245502 Certificate TypeCertificate of Analysis Date13/11/2023 Catalog NumberA18481.0F,A18481.AE,A18481.AP
Lot Number10235936 Certificate TypeCertificate of Analysis Date13/11/2023 Catalog NumberA18481.0F,A18481.AE,A18481.AP
Lot Number10235935 Certificate TypeCertificate of Analysis Date13/10/2023 Catalog NumberA18481.0F,A18481.AE,A18481.AP
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Styrene
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 3
  • Aspiration toxicity Category 1
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 3
  • Reproductive toxicity Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 1
  • Specific target organ toxicity Category 3
Hazard Statement
  • H226-Flammable liquid and vapour.
  • H304-May be fatal if swallowed and enters airways.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H332-Harmful if inhaled.
  • H335-May cause respiratory irritation.
  • H361d-Suspected of damaging the unborn child.
  • H372-Causes damage to organs.
  • H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P331-Do NOT induce vomiting.
Supplemental information
  • MIXTURE LIST-Contains : Styrene

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Styrene, 99%, stab. with 10-15ppm 4-tert-butylcatechol, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.