missing translation for 'onlineSavingsMsg'
Learn More
Succinic Acid 99%, Thermo Scientific Chemicals Product Code.: 10144243

Succinic Acid 99%, Thermo Scientific Chemicals

Product Code. 10144243
1 kg, Plastic bottle
Click to view available options
Quantity:
250 g
500 g
1 kg
2.5 kg
10 kg
Packaging:
Plastic bottle
Plastic drum
This item is not returnable. View return policy

Product Code. 10144243

Brand: Thermo Scientific Acros 158740010

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 110-15-6
Molecular Formula C4H6O4
Molecular Weight (g/mol) 118.09
MDL Number MFCD00002789
InChI Key KDYFGRWQOYBRFD-UHFFFAOYSA-N
Synonym succinic acid, amber acid, asuccin, dihydrofumaric acid, bernsteinsaure, katasuccin, wormwood acid, succinate, ethylenesuccinic acid, 1,2-ethanedicarboxylic acidShow MoreShow Less
PubChem CID 1110
ChEBI CHEBI:15741
IUPAC Name butanedioic acid
SMILES OC(=O)CCC(O)=O

Specifications

Melting Point 185.0°C to 190.0°C
Boiling Point 235.0°C
Assay Percent Range 99%
Linear Formula HO2CCH2CH2CO2H
Solubility Information Solubility in water: 80g/L (20°C). Other solubilities: 1 kg/l boiling water, 1 g/18.5 mL alcohol - 1 g/6.3 mL methanol, 1 g/36 mL acetone - 1 g/20 mL glycerol, practically insoluble in petroleum ether, benzene,, carbon disulfide and carbon tetrachloride
Formula Weight 118.09
Percent Purity 99%
Quantity 1 kg
Flash Point 206°C
Packaging Plastic bottle
Chemical Name or Material Succinic acid, 99%
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0466234 Certificate TypeCertificate of Analysis Date25/09/2024 Catalog Number158740100,158740010,158740025,158742500,158745000
Lot NumberA0461211 Certificate TypeCertificate of Analysis Date20/03/2024 Catalog Number158740100,158740010,158740025,158742500,158745000
Lot NumberA0450263 Certificate TypeCertificate of Analysis Date10/02/2023 Catalog Number158740100,158740010,158740025,158742500,158745000
Lot NumberA0394478 Certificate TypeCertificate of Analysis Date15/12/2022 Catalog Number158740100,158740010,158740025,158742500,158745000
Lot NumberA0397958 Certificate TypeCertificate of Analysis Date14/12/2022 Catalog Number158740100,158740010,158740025,158742500,158745000
compliance-icons
Product Identifier
  • Succinic acid
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
Hazard Statement
  • H318-Causes serious eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Succinic Acid 99%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.