Learn More
Mettler Toledo™ Sulfide Half Cell Electrode
Brand:  Mettler Toledo™ 51107675
1105.00 GBP valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Product Code. 11937701
  • £1150.00 / Each
In Stock
Explore more special offers
Add to basket
Specifications
Solid State Membrane
0°C to 80°C
4 to 13
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Mettler Toledo™ Sulfide Half Cell Electrode > Membrane: Solid State